Learn More
CTL Scientific Supply Corp NANOCOLOR ortho- and total-Phosphate 5, contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985081
Encompass_Preferred
Tube test for the determination of ortho- and total Phosphate. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Phosphate (ortho) - 0.5–15.0 mg/L PO₄³⁻ , Phosphate (ortho) - 0.20–5.00 mg/L P , Phosphate (total) - 0.5–15.0 mg/L PO₄³⁻ , Phosphate (total) - 0.20–5.00 mg/L PManufactured by Macherey-Nagel
Catalog No. NC2958181