Learn More
CTL Scientific Supply Corp NANOCOLOR ortho and total Phosphate 50, contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985079
Encompass_Preferred
Tube test for the determination of ortho- and total Phosphate. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Phosphate (ortho) - 10.0–50.0 mg/L P, Phosphate (ortho) - 30–150 mg/L PO₄³⁻ , Phosphate (total) - 10.0–50.0 mg/L P, Phosphate (total) - 30–150 mg/L PO₄³⁻ *This item is hazardous and cannot ship Parcel Post. It is required to ship UPS Ground*Manufactured by Macherey-Nagel
Catalog No. NC2958203