Learn More
Proteintech Group Inc Nano-Secondary alpaca anti-mouse IgG1, recombinant VHH, Alexa Fluor 568 [CTK0103, CTK0104]
SDP
Supplier:  Proteintech Group Inc SMS1AF568110
Encompass
Nano-Secondary alpaca anti-mouse IgG1, Fc-specific recombinant VHH reagent uses a novel class of anti-mouse IgG isotype specific antibodies. This secondary antibody product consists of Nanobodies that bind to mouse IgG1 with high affinity & specificity.
Catalog No. 50-225-8830