Learn More
Proteintech Group Inc Nano-Secondary alpaca anti-mouse IgG1, recombinant VHH, Alexa Fluor 488 [CTK0103, CTK0104]
SDP
Catalog No. 502258829 Shop All Proteintech Group Inc Products
Encompass
Nano-Secondary alpaca anti-mouse IgG1, Fc-specific recombinant VHH reagent uses a novel class of anti-mouse IgG isotype specific antibodies. This secondary antibody product consists of Nanobodies that bind to mouse IgG1 with high affinity & specificity.
This item is not returnable. View return policy
Catalog No. 50-225-8829 Supplier Proteintech Group Inc Supplier No. SMS1AF4881100
Nano-Secondary alpaca anti-mouse IgG1, Fc-specific recombinant VHH reagent uses a novel class of anti-mouse IgG isotype specific antibodies. This secondary antibody product consists of Nanobodies that bind to mouse IgG1 with high affinity & specificity.