Learn More
Proteintech Group Inc Nano-Secondary alpaca anti-mouse IgG1, recombinant VHH, for 2x Cys conjugation [CTK0103]
SDP
Catalog No. 502258846 Shop All Proteintech Group Inc Products
Encompass
Monovalent, recombinant secondary single domain antibody to mouse IgG1: alpaca monoclonal Nanobody, Fc-specific, for cysteine conjugation with thiol-reactive reagents, e.g. maleimides
This item is not returnable. View return policy
Catalog No. 50-225-8846 Supplier Proteintech Group Inc Supplier No. SMSG1CYS21500
Monovalent, recombinant secondary single domain antibody to mouse IgG1: alpaca monoclonal Nanobody, Fc-specific, for cysteine conjugation with thiol-reactive reagents, e.g. maleimides