Learn More
LUNA NANOTECH INC Nano Silica Spin Columns, 200 Columns
Supplier:  LUNA NANOTECH INC USP002200N
Encompass_Preferred
PuroSPIN™ NANO Spin Columns for DNA / RNA Extraction and Purification, 200 columns
Have you ever ran out of columns that come with your favorite nucleic acid extraction kit but still had a lot of left-over buffer reagents remaining? Instead of buying a new expensive kit you can use PuroSPIN™ columns with the left-over buffers as a cost-effective alternative. Simply use PuroSPIN™ columns while following the manufacturer-specific protocols.
PuroSPIN™ NANO silica spin columns bind both DNA and RNA
Low 6 μL elution volume - maximizes concentration of purified product
5 μg nucleic acid binding capacity
Compatible as cost-effective replacement columns for nucleic acid extraction kits from Qiagen, ThermoFisher, Bio-Rad, Promega and other manufacturers
Each item includes both the spin column and a 2 mL sample collection tube
Extraction of DNA and RNA from bacteria, viruses, yea
Catalog No. 50-256-6912