Learn More
Waters Corp Waters Nano-Tee, .006 in. Thru, UPLC, nanoACQUITY System
Catalog No. NC1734751 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This nano-tee assembly is designed for use with the nanoACQUITY UPLC system. It features a .006 in. thru-bore and is compatible with various fluidic configurations, providing essential connectivity for high-performance liquid chromatography applications.
  • Designed for 10,000 psi high-performance liquid chromatography systems
  • Compatible with 2D with dilution fluidic configuration
  • Compatible with direct injection fluidic configuration
  • Compatible with single pump fluidic configuration
  • Compatible with vented dual pump trapping fluidic configuration
  • Functions as an essential fitting for system components
Catalog No. NC1734751 Supplier Waters Corp Supplier No. 289002576
This nano-tee assembly is designed for use with the nanoACQUITY UPLC system. It features a .006 in. thru-bore and is compatible with various fluidic configurations, providing essential connectivity for high-performance liquid chromatography applications.
  • Designed for 10,000 psi high-performance liquid chromatography systems
  • Compatible with 2D with dilution fluidic configuration
  • Compatible with direct injection fluidic configuration
  • Compatible with single pump fluidic configuration
  • Compatible with vented dual pump trapping fluidic configuration
  • Functions as an essential fitting for system components