Learn More
CTL Scientific Supply Corp NANOCOLOR total Nitrogen TNb 22, contains 20 tube tests
SDP
Catalog No. NC2958191 Shop All CTL Scientific Supply Products
Encompass_Preferred
Tube test for the determination of Total Nitrogen with the easiest handling available. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Total Nitrogen - 0.5–22.0 mg/L N *This item is hazardous and cannot ship Parcel Post. It is required to ship UPS Ground*Manufactured by Macherey-Nagel
Catalog No. NC2958191 Supplier CTL Scientific Supply Corp Supplier No. 985083
Tube test for the determination of Total Nitrogen with the easiest handling available. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Total Nitrogen - 0.5–22.0 mg/L N *This item is hazardous and cannot ship Parcel Post. It is required to ship UPS Ground*Manufactured by Macherey-Nagel