Learn More
Optimize Technologies EXP™2 Nano Trap In-Line Holder Kit with 6-40 Threads (Includes 1/32" PEEKsil tubing and 4 EXP™2 Fittings)
SDP
Catalog No. 502006352
Encompass_Preferred
The EXP™2 Nano Trap System provides the finest low-volume hardware and connections to minimize extra column effects and sample dispersion. The packed beds are extremely versatile: robust for applications calling for trapping in one direction followed by elution in the reverse while adding the absolute lowest swept volume for vented-trap applications where the trap is loaded and eluted in the same direction. EXP™2 Nano Trap Holders are available with 10-32 threaded connections for 1/16” tube ports or 6-40 threaded connections for 1/32” tube ports. Multiple bed volumes and bonded phases are available. EXP™2 fittings allow the Nano Trap to achieve the highest performance at UHPLC pressures while maintaining a small profile to fit tight spaces. Couple with narrow bore (25um, 50um, 100um) PEEKsil tubing to deliver top performance. Additionally, the product features a low swept volume design for peak sharpness and is rated to 20,000+psi.
Catalog No. 50-200-6352 Supplier Optimize Technologies Supplier No. 150205088
The EXP™2 Nano Trap System provides the finest low-volume hardware and connections to minimize extra column effects and sample dispersion. The packed beds are extremely versatile: robust for applications calling for trapping in one direction followed by elution in the reverse while adding the absolute lowest swept volume for vented-trap applications where the trap is loaded and eluted in the same direction. EXP™2 Nano Trap Holders are available with 10-32 threaded connections for 1/16” tube ports or 6-40 threaded connections for 1/32” tube ports. Multiple bed volumes and bonded phases are available. EXP™2 fittings allow the Nano Trap to achieve the highest performance at UHPLC pressures while maintaining a small profile to fit tight spaces. Couple with narrow bore (25um, 50um, 100um) PEEKsil tubing to deliver top performance. Additionally, the product features a low swept volume design for peak sharpness and is rated to 20,000+psi.