Learn More
CTL Scientific Supply Corp NANOCOLOR Aluminium 07, contains 19 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985098
Encompass_Preferred
Tube test for the determination of Aluminum. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Aluminum - 0.02–0.70 mg/L Al³⁺Manufactured by Macherey-Nagel
Catalog No. NC2958035