Learn More
CTL Scientific Supply Corp NANOCOLOR Ammonium 200, contains 20 tube tests
SDP
Catalog No. NC2958107 Shop All CTL Scientific Supply Products
Encompass_Preferred
Tube test for the determination of Ammonium. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Ammonium - 30–160 mg/L NH₄-N, Ammonium - 40–200 mg/L NH₄⁺Manufactured by Macherey-Nagel
Catalog No. NC2958107 Supplier CTL Scientific Supply Corp Supplier No. 985006
Tube test for the determination of Ammonium. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Ammonium - 30–160 mg/L NH₄-N, Ammonium - 40–200 mg/L NH₄⁺Manufactured by Macherey-Nagel