Learn More
CTL Scientific Supply Corp NANOCOLOR Ammonium 3, contains 20 tube tests
SDP
Catalog No. NC2958110 Shop All CTL Scientific Supply Products
Encompass_Preferred
Tube test for the determination of Ammonium. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Ammonium - 0.04–2.30 mg/L NH₄-N, Ammonium - 0.05–3.00 mg/L NH₄⁺Manufactured by Macherey-Nagel
Catalog No. NC2958110 Supplier CTL Scientific Supply Corp Supplier No. 985003
Tube test for the determination of Ammonium. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Ammonium - 0.04–2.30 mg/L NH₄-N, Ammonium - 0.05–3.00 mg/L NH₄⁺Manufactured by Macherey-Nagel