Learn More
CTL Scientific Supply Corp NANOCOLOR BOD₅-TT, contains 22 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985825
Encompass_Preferred
Tube test for the determination of BOD₅ in test tubes. Precise rapid tests for all kind of water and waste water analysis. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: BOD5 - 0.5–3000 mg/L O2 - *This item is hazardous and cannot ship Parcel Post. It is required to ship UPS Ground*Manufactured by Macherey-Nagel
Catalog No. NC2958117