Learn More
CTL Scientific Supply Corp NANOCOLOR Chlorine/Ozone 2, contains 20 tube tests
SDP
Catalog No. NC2958118 Shop All CTL Scientific Supply Products
Encompass_Preferred
Tube test for the determination of Chlorine and Ozone. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Chlorine (free)-0.05–2.50 mg/L Cl₂, Chlorine (total)-0.05–2.50 mg/L Cl₂, Ozone-0.05–2.00 mg/L O₃Manufactured by Macherey-Nagel
Catalog No. NC2958118 Supplier CTL Scientific Supply Corp Supplier No. 985017
Tube test for the determination of Chlorine and Ozone. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Chlorine (free)-0.05–2.50 mg/L Cl₂, Chlorine (total)-0.05–2.50 mg/L Cl₂, Ozone-0.05–2.00 mg/L O₃Manufactured by Macherey-Nagel