Learn More
CTL Scientific Supply Corp NANOCOLOR COD 40, contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985027
Encompass_Preferred
Tube test for the determination of COD. Precise rapid tests for all kind of water and waste water samples according to ISO 15705. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: COD - 2–40 mg/L O₂ *This item is hazardous and cannot ship Parcel Post. It is required to ship UPS Ground* REFRIGERATED ITEMManufactured by Macherey-Nagel
Catalog No. NC2958149