Learn More
CTL Scientific Supply Corp NANOCOLOR Nitrate 8, contains 20 tube tests
SDP
Catalog No. NC2958197 Shop All CTL Scientific Supply Products
Encompass_Preferred
Tube test for the determination of Nitrate. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Nitrate - 0.30–8.00 mg/L NO₃-N, Nitrate - 1.3–35.0 mg/L NO₃⁻ *This item is hazardous and cannot ship Parcel Post. It is required to ship UPS Ground*Manufactured by Macherey-Nagel
Catalog No. NC2958197 Supplier CTL Scientific Supply Corp Supplier No. 985065
Tube test for the determination of Nitrate. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Nitrate - 0.30–8.00 mg/L NO₃-N, Nitrate - 1.3–35.0 mg/L NO₃⁻ *This item is hazardous and cannot ship Parcel Post. It is required to ship UPS Ground*Manufactured by Macherey-Nagel