Learn More
CTL Scientific Supply Corp NANOCOLOR Sulfate MR 400, contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985060
Encompass_Preferred
Tube test for the determination of Sulfate. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Sulfate - 40-400 mg/L SO₄²⁻Manufactured by Macherey-Nagel
Catalog No. NC2958201