Learn More
CTL Scientific Supply Corp NANOCOLOR Zinc 4, contains 20 tube tests
SDP
Supplier:  CTL Scientific Supply Corp 985096
Encompass_Preferred
Tube test for the determination of Zinc. Precise rapid tests for all kind of water and waste water samples. Time-saving and reliable analysis together with Macherey-Nagel NANOCOLOR photometers. Measuring range: Zinc - 0.10–4.00 mg/L Zn²⁺ *This item is hazardous and cannot ship Parcel Post. It is required to ship UPS Ground*Manufactured by Macherey-Nagel
Catalog No. NC2958195