Learn More
Electron Microscopy Sciences Rubis Nano Tweezers Style 3, 120 mm OAL
SDP
Catalog No. 5024053
Encompass_Preferred
Ultra-modern nanotechnology closes even the tiniest of pores on the surface and this innovation has resulted in the cleanest tweezers surface ever produced. Made from the  finest stainless steel grade, the Lotus effect repels dirt, oil, water and other liquids. These tweezers are a perfect companion in the clean room or any sensitive laboratory areas where contaminant free tools is required.
OAL: 120 mm
Catalog No. 50-240-53 Supplier Electron Microscopy Sciences Supplier No. 781483
Ultra-modern nanotechnology closes even the tiniest of pores on the surface and this innovation has resulted in the cleanest tweezers surface ever produced. Made from the  finest stainless steel grade, the Lotus effect repels dirt, oil, water and other liquids. These tweezers are a perfect companion in the clean room or any sensitive laboratory areas where contaminant free tools is required.
OAL: 120 mm