Learn More
Ted Pella Inc Nano-W™Negative Stain, 5ml
SDP
Catalog No. NC1646615
Encompass
Originally intended for staining samples with gold nanoparticles, these convenient ready-to-use pre-made negative stains have been since utilized for many other applications, including pre-screening samples for cryo-EM. NanoVan™is based on vanadium and thus gives lighter staining, for best visibility of gold and silver nanoparticles in the sample. Nano-W is tungsten-based and recommended for those cases when higher contrast is desired; it is comparable to PTA negative stain.
This item is not returnable. View return policy
Catalog No. NC1646615 Supplier Ted Pella Inc Supplier No. 14702
Originally intended for staining samples with gold nanoparticles, these convenient ready-to-use pre-made negative stains have been since utilized for many other applications, including pre-screening samples for cryo-EM. NanoVan™is based on vanadium and thus gives lighter staining, for best visibility of gold and silver nanoparticles in the sample. Nano-W is tungsten-based and recommended for those cases when higher contrast is desired; it is comparable to PTA negative stain.