Learn More
Waters Corp NANOACQUITY HSS T3, 1.8 UM, 75
Supplier:  Waters Corp 186005777
Encompass_Preferred
nanoACQUITY HSS T3, 1.8 um, 75 um x 200 mm,Reversed-Phase,Chemistry: C18,Particle Substrate: Silica,USP: L1,Pore Size: 100A,pH Range: 2 - 8
Catalog No. 50-466-261