Learn More
Waters Corp Waters Sample Manager Performance Maintenance Kit, nanoACQUITY, Annual, 5000/10000 psi Systems
Catalog No. 50791579 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit contains all the necessary parts to keep the sample manager operating at peak performance annually. It is designed to fit into both 5000 psi and 10,000 psi nanoACQUITY systems.
  • Air chassis filter
  • Injector outlet tubes
  • Needle
  • System outlet tubes
  • Inject valve pod
  • Wash syringe
  • Injector syringe
  • Sample loop
  • O-ring
  • Vent valve cartridge
Catalog No. 50-791-579 Supplier Waters Corp Supplier No. 201000182
Add to Cart
Add to Cart
Request a Quote
This kit contains all the necessary parts to keep the sample manager operating at peak performance annually. It is designed to fit into both 5000 psi and 10,000 psi nanoACQUITY systems.
  • Air chassis filter
  • Injector outlet tubes
  • Needle
  • System outlet tubes
  • Inject valve pod
  • Wash syringe
  • Injector syringe
  • Sample loop
  • O-ring
  • Vent valve cartridge