Learn More
Waters Corp Waters Atlantis dC18 nanoACQUITY UPLC Column, 100 μm I.D., 100 mm, 3 μm Particle
Catalog No. 50781808 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This nanoACQUITY UPLC Column is engineered for high-performance liquid chromatography applications. It utilizes the Atlantis dC18 stationary phase and precise dimensions to deliver efficient and reliable chromatographic separations.
  • Atlantis dC18 stationary phase for optimal separation
  • 100 μm inner diameter
  • 100 mm length
  • 3 μm particle size for efficient chromatographic performance
  • Designed for use with UPLC systems rated to 10,000 psi
Catalog No. 50-781-808 Supplier Waters Corp Supplier No. 186003501
This nanoACQUITY UPLC Column is engineered for high-performance liquid chromatography applications. It utilizes the Atlantis dC18 stationary phase and precise dimensions to deliver efficient and reliable chromatographic separations.
  • Atlantis dC18 stationary phase for optimal separation
  • 100 μm inner diameter
  • 100 mm length
  • 3 μm particle size for efficient chromatographic performance
  • Designed for use with UPLC systems rated to 10,000 psi