Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation UPLC Column, BEH130 C18, 75 μm ID, 1.7 μm, 100 mm

Catalog No. 50781831 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This UPLC column is part of the Peptide Separation Technology nanoACQUITY UPLC Columns series and is designed for use with nanoACQUITY UPLC systems rated to 10,000 psi only.

  • Features BEH130 C18 chemistry.
  • Has an inner diameter of 75 μm.
  • Has a length of 100 mm.
  • Particle size of 1.7 μm.
  • Designed for peptide separation technology.
  • Compatible with UPLC systems rated to 10,000 psi.

Catalog No. 50-781-831 Supplier Waters Corp Supplier No. 186003542
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This UPLC column is part of the Peptide Separation Technology nanoACQUITY UPLC Columns series and is designed for use with nanoACQUITY UPLC systems rated to 10,000 psi only.

  • Features BEH130 C18 chemistry.
  • Has an inner diameter of 75 μm.
  • Has a length of 100 mm.
  • Particle size of 1.7 μm.
  • Designed for peptide separation technology.
  • Compatible with UPLC systems rated to 10,000 psi.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.