Learn More
Waters Corp NANOACQUITY UPLC BEH C18 COLUM
Supplier:  Waters Corp 186003544
Encompass_Preferred
nanoACQUITY UPLC BEH C18 Column, 1.7 um, 75 um x 200 mm, 10K psi,Reversed-Phase,Chemistry: C18,Particle Substrate: Hybrid,USP: L1,Pore Size: 130A,pH Range: 1 - 12
Catalog No. 50-781-833