Learn More
Waters Corp ACQUITY UPLC M-Class Peptide BEH C18 Column, 1.7 μm, 75 μm × 150 mm
Supplier: Waters Corp 186003543
- Acuity UPLC M-Class Peptide BEH C18 Column
- BEH particle type
- Pore size: 130Å
- Particle size: 1.7 μm
- Inner diameter: 75 μm
- Length: 150 mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.