Learn More
Waters Corp WATERS CORPORATION nanoACQUITY UPLC Column, BEH300 C18, 75 μm ID, 250 mm Length
Catalog No. 50782036 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The nanoACQUITY UPLC BEH300 C18 column is part of the Peptide Separation Technology nanoACQUITY UPLC Columns series for nanoACQUITY UPLC systems rated to 10,000 psi. These columns are manufactured to exacting specifications, providing peak symmetry for maximum sensitivity and accurate quantitation. They are fritted at both ends to withstand pressure changes, offering superior longevity. Each column is individually tested to ensure stringent quality control.
  • 75 μm inner diameter
  • 250 mm length
  • 1.7 μm particle size
  • Designed for UPLC systems rated to 10,000 psi
  • Manufactured to exacting specifications
  • Outstanding peak symmetry for maximum sensitivity and accurate quantitation
  • Fritted at both ends to withstand pressure changes, offering superior longevity
  • Individually tested for stringent quality control
Catalog No. 50-782-036 Supplier Waters Corp Supplier No. 186003815
The nanoACQUITY UPLC BEH300 C18 column is part of the Peptide Separation Technology nanoACQUITY UPLC Columns series for nanoACQUITY UPLC systems rated to 10,000 psi. These columns are manufactured to exacting specifications, providing peak symmetry for maximum sensitivity and accurate quantitation. They are fritted at both ends to withstand pressure changes, offering superior longevity. Each column is individually tested to ensure stringent quality control.
  • 75 μm inner diameter
  • 250 mm length
  • 1.7 μm particle size
  • Designed for UPLC systems rated to 10,000 psi
  • Manufactured to exacting specifications
  • Outstanding peak symmetry for maximum sensitivity and accurate quantitation
  • Fritted at both ends to withstand pressure changes, offering superior longevity
  • Individually tested for stringent quality control