Learn More
Waters Corp nanoACQUITY UPLC BEH300 C18, 150 μm ID, 100 mm Length
Catalog No. 50782033 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The nanoACQUITY UPLC BEH300 C18 column is designed for peptide separation technology, offering high performance for demanding applications. It features a 150 μm inner diameter, 100 mm length, and 1.7 μm particle size.
  • Compatible with UPLC systems rated to 10,000 psi.
  • Not for use with UPLC systems rated to 5,000 psi.
  • Part of peptide separation technology product line.
Catalog No. 50-782-033 Supplier Waters Corp Supplier No. 186003812
Add to Cart
Add to Cart
The nanoACQUITY UPLC BEH300 C18 column is designed for peptide separation technology, offering high performance for demanding applications. It features a 150 μm inner diameter, 100 mm length, and 1.7 μm particle size.
  • Compatible with UPLC systems rated to 10,000 psi.
  • Not for use with UPLC systems rated to 5,000 psi.
  • Part of peptide separation technology product line.