Learn More
Polysciences, Inc. Nanobead NIST Traceable Particle Size Standard, 400nm | 15mL
SDP
Catalog No. NC1665480
Encompass
Precision Size Standards are measured on in-house instruments calibrated with NIST (National Institute of Standards and Technology) Standard Reference Materials. Each product is supplied with a Certificate of Analysis documenting traceability to NIST Standard Reference Materials. The size, standard deviation, coefficient of variation, and lot number are printed on the label for easy reference.
Catalog No. NC1665480 Supplier Polysciences, Inc. Supplier No. 6401715
Precision Size Standards are measured on in-house instruments calibrated with NIST (National Institute of Standards and Technology) Standard Reference Materials. Each product is supplied with a Certificate of Analysis documenting traceability to NIST Standard Reference Materials. The size, standard deviation, coefficient of variation, and lot number are printed on the label for easy reference.