Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nanoclay, hydrophilic bentonite

Catalog No. 682659500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-265-9500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-265-9500G Supplier Sigma Aldrich Supplier No. 682659500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Untreated (no organic modification) hydrophilic clay for dispersion in water-based polymers and coatingsRheology controlIncrease barrierEnhance chemical resistanceNanomer(R) clays are products of Nanocor CorporationProduct of Nanocor(R), Inc.

Specifications

CAS 1302-78-9
MDL Number MFCD00132796
Quantity 500 g
Synonym Nanomer™ PGV; Nanomer™ clay; Montmorillinite clay, bentonite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.