Learn More
Thermo Scientific™ NanoDrop™ Eight IQ/OQ Kits
Catalog No. 13400535 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
ND Eight Spec IQ/OQ - A4
ND Eight Spec IQ/OQ - US
ND Eight Spec IQ/OQ Requal
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
13-400-535 ND Eight Spec IQ/OQ - A4
13-400-534 ND Eight Spec IQ/OQ - US
13-400-536 ND Eight Spec IQ/OQ Requal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-400-535 Supplier Thermo Scientific™ Supplier No. 840371300
Add to Cart
Edge
Add to Cart
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ). Certified standards for verifying instrument performance to factory specifications are available separately.
Quantify your samples with confidence using the NanoDrop Eight multichannel spectrophotometer. This is the next generation NanoDrop 8000 with improved hardware and the best software features from the Thermo Scientifict™NanoDrop™ One/OneC spectrophotometer.
Acclaro Contaminant Identification ensures samples are high quality
Know true concentration of your nucleic acid or protein without the interfering effects of sample contaminants. Understanding your sample composition helps downstream experiments go right the first time.
• Contaminant identification – recognizes common contaminants in nucleic acid and protein samples that could inhibit downstream reactions or falsely elevate calculated concentrations
• Corrected calculations – applies chemometric algorithms to determine the true concentration of your analyte without the contaminant
• Differentiate DNA from RNA – algorithms can identify mammalian RNA contaminating a dsDNA sample or mammalian dsDNA contaminating an RNA sample
NanoDrop Eight enhancements
• Wider dynamic range – measure higher concentration samples without dilution – up to 10,000 ng/μL dsDNA or 145 mg/mL IgG
• Auto-Measure and Auto-Blank functions – multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down. These functions can be turned ON or OFF
• Ergonomic ambidextrous pipetting – Pedestal array is aligned perpendicular to the front of the instrument creating a natural pipetting motion for users; the pipette guide is magnetic and
movable from side-to-side allowing left- and righthanded users to quickly and comfortably dispense samples
• Automated exports for LIMS – set a directory for data to be automatically exported; have your LIMS software sweep the folder to pull in NanoDrop Eight instrument data for archiving
Regulatory compliance made easy
Optional Thermo Scientific™ SciVault™ Software helps your lab stay compliant with federal data
regulations every step of the way.
• Streamline management – Optional SciVault Software integrates into NanoDrop Eight Software user interface
• Comply with US FDA 21 CFR Part 11 – Control user account access, apply digital signatures, and review audit logs
• Network multiple instruments – Install SciVault Software on a central computer to control privileges and view audit logs across multiple NanoDrop Eight Instruments in different labs
• Validate your systems – IQ/OQ documents and services available to confirm your instrument is running within manufacturer’s specifications
Software updates to make your time more productive
• Ready to start – NanoDrop Eight Software comes hard-coded with the most frequently used measurement applications including dsDNA, RNA, Protein A280, and UV-Vis
• Always improving – user input keeps NanoDrop Software constantly moving forward with updates that deliver greater capabilities and efficiencies in the lab. Visit thermoscientific.com/nanodropsw to see and download the latest software applications. Register your instrument there as well so you won’t miss any enhancements.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) NanoDrop Eight Spectrophotometers
Product Type ND Eight Spec IQ/OQ - A4
Includes Qualification Kit includes: IQ/OQ procedure for NanoDrop Eight Spectrophotometers, Qualification Binder Kit (A4 Size), CHEM-PR1-KIT, CHEM-PV-8
Which is the best DNA or RNA quantification instrument?
All of the DNA instruments offered by Thermo Fisher Scientific are of excellent quality. To find the right instrument to quantify DNA or RNA in your lab, explore the detailed comparison of UV-Vis spectrophotometers and fluorometers. Things like sensitivity, throughput and budget may be initial considerations when selecting a DNA quantification instrument. You may also consider whether you need target specificity, sample purity information, RNA quality information, or broad dynamic range.
What is the difference between UV and fluorescence DNA quantification? Which DNA quantification technology should I choose?
UV and fluorescence technologies work differently to quantify DNA. UV quantification relies on the intrinsic absorptivity of DNA and RNA molecules, while fluorescence quantification uses dyes that specifically bind to your molecule of choice. With UV technology, quantification isn't as sensitive, but it has a broader dynamic range and also gives data about sample purity‒plus it is faster because there is no reagent prep. With fluorescence technology you get higher sensitivity and molecule-specific data, but it has lower dynamic range and reagent prep is required. The technology you need will depend on what features are important for your lab and experimentation.