Learn More
Thermo Scientific™ NanoDrop™ Eight Pipette Guide
Catalog No. 13400537 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
NanoDrop Eight Spectrophotometers
Catalog No. For Use With (Equipment)
13-400-537 NanoDrop Eight Spectrophotometers
1 options
Catalog No. 13-400-537 Supplier Thermo Scientific™ Supplier No. 840343600
Add to Cart
Add to Cart
The Thermo Scientific™ NanoDrop™ Eight Pipette guide allows you to comfortably and accurately pipette samples onto all eight wells of the instrument using an 8-channel precision pipettor.
The Thermo Scientific™ NanoDrop™ Eight Pipette guide allows you to comfortably and accurately pipette samples onto all eight wells of the instrument using an 8-channel precision pipettor.
The pipette guide is magnetic and movable from side-to-side allowing left- and right-handed users to quickly and comfortably dispense samples
TRUSTED_SUSTAINABILITY
Specifications
Description NanoDrop Eight Pipette Guide
For Use With (Equipment) NanoDrop Eight Spectrophotometers
Which is the best DNA or RNA quantification instrument?
All of the DNA instruments offered by Thermo Fisher Scientific are of excellent quality. To find the right instrument to quantify DNA or RNA in your lab, explore the detailed comparison of UV-Vis spectrophotometers and fluorometers. Things like sensitivity, throughput and budget may be initial considerations when selecting a DNA quantification instrument. You may also consider whether you need target specificity, sample purity information, RNA quality information, or broad dynamic range.
What is the difference between UV and fluorescence DNA quantification? Which DNA quantification technology should I choose?
UV and fluorescence technologies work differently to quantify DNA. UV quantification relies on the intrinsic absorptivity of DNA and RNA molecules, while fluorescence quantification uses dyes that specifically bind to your molecule of choice. With UV technology, quantification isn't as sensitive, but it has a broader dynamic range and also gives data about sample purity‒plus it is faster because there is no reagent prep. With fluorescence technology you get higher sensitivity and molecule-specific data, but it has lower dynamic range and reagent prep is required. The technology you need will depend on what features are important for your lab and experimentation.