Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NanoDrop™ Eight Spectrophotometer and Qubit™ Flex Fluorometer PROMO

Catalog No. NDEQBFLXP26 Shop All Thermo Scientific Products
Change view
Click to view available options
Display:
Touchscreen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display
NDEQBFLXP26 Touchscreen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NDEQBFLXP26 Supplier Thermo Scientific™ Supplier No. NDEQBFLEX2PM26
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enhance throughput of your UV-Vis absorbance and fluorescence measurements with this instrument bundle. Measure dsDNA, RNA, protein, or more, 8-samples at a time.

The Thermo Scientific™ NanoDrop™ Eight Multichannel UV-Vis spectrophotometer measures 8 samples at a time in less than 20 seconds, enabling quick and efficient analysis of your sample quantity and quality. Built-in Thermo Scientific™ Acclaro™ Sample Intelligence technology can identify sample contaminants and report true sample concentrations. The Qubit Flex Fluorometer is a benchtop fluorometer designed for highly accurate quantification of DNA, RNA, microRNA, and protein. With the Qubit Flex Fluorometer you have the flexibility to directly measure fluorescence of up to eight samples simultaneously.

NanoDrop Eight spectrophotometer features

  • Wider dynamic range - measure higher concentration samples without dilution: up to 10,000 ng/μL dsDNA or 145 mg/mL IgG
  • Auto-Measure and Auto-Blank functions - multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down; these functions can be turned ON or OFF
  • Ergonomic ambidextrous pipetting - the pedestal array is aligned perpendicular to the front of the instrument creating a natural pipetting motion for users; the pipette guide is magnetic and movable from side-to-side allowing left- and righthanded users to quickly and comfortably dispense samples
  • Automated exports for LIMS - data can be automatically exported to a selected directory; LIMS software can sweep the folder to pull in NanoDrop Eight instrument data for archiving

Acclaro Contaminant Identification ensures samples are high quality

Specialized software algorithms provide clear information about the true concentration of your nucleic acid or protein despite any possible interfering effects of sample contaminants, so you can understand your sample composition and ensure your downstream experiments go right the first time.

  • Contaminant identification - recognizes common contaminants in nucleic acid and protein samples that could inhibit downstream reactions or falsely elevate calculated concentrations
  • Corrected calculations - applies chemometric algorithms to determine the true concentration of your analyte without the contaminant
  • DNA and RNA differentiation - identifies mammalian RNA contaminating a dsDNA sample or mammalian dsDNA contaminating an RNA sample

Qubit Flex Fluorometer features

  • Improved throughput - measures multiple samples and quantify DNA, RNA, and protein up to 50% faster than single-sample readers
  • Flexibility - measures up to eight samples simultaneously
  • Complete optimized system - works seamlessly with Qubit reagents to generate reliable, sensitive, and specific results
  • Accurate quantification - delivers high levels of accuracy using only 1 to 20 μL of sample, even with very dilute samples
  • On-board calculators - quickly generates Qubit working solution preparation instructions and determines the molarity of your samples
  • Easy to use - a large 8-inch, state-of-the-art color touchscreen and intuitive user interface results in easy workflow navigation
  • Data export flexibility - data is transferred easily using the included WiFi dongle, a USB drive, or directly to your computer
  • Several languages available - choices include English, French, Spanish, Italian, German, simplified Chinese, and Japanese
  • Verification test - a pre-loaded program is included to verify performance of the instrument
  • Qubit Flex SmartStart Digital Educational Experience - includes interactive visual demonstrations of how to install, operate, and maintain your instrument
  • Three-year Extended Warranty - if your instrument needs repair we will send you a factory-certified refurbished replacement
TRUSTED_SUSTAINABILITY

Specifications

Display Touchscreen
Product Line NanoDrop
Type Spectrophotometer with Fluorometer
For Use With (Application) Sample Concentration
Includes ND Eight and QBFlex
Storage Requirements Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.