Learn More
Thermo Scientific™ NanoDrop™ One/OneC Productivity Kits
Description
The Thermo Scientific™ NanoDrop™ One and NanoDrop™ OneC Productivity Kits allow you to improve the productivity of your NanoDrop One and NanoDrop OneC Microvolume UV-Vis Spectrophotometers.
The NanoDrop One Productivity Kit includes:
- NanoDrop-branded Finnpipette™ F1 Adjustable-Volume Pipette , 0.2–2 μL
- Microfiber screen wipe for NanoDrop One Spectrophotometer
- NanoDrop PR-1 Reconditioning Compound Kit
- NanoDrop PV-1 Performance Verification Solution
- I love my NanoDrop sticker
The NanoDrop OneC Productivity Kit includes:
- NanoDrop-branded Finnpipette™ F1 Adjustable Volume Pipette , 0.2–2μL
- Microfiber screen wipe for NanoDrop OneC spectrophotometer
- NanoDrop PR-1 Reconditioning Compound Kit
- NanoDrop PV-1 Performance Verification Solution
- 10mm path quartz cuvette
- Cuvette stir bars, 5x2mm, pack of 10
- I love my NanoDrop sticker
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
| Type | NanoDrop One Productivity Kit |
| For Use With (Equipment) | NanoDrop One |
| Includes |
|
| Storage Requirements | Store at room temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.