Learn More
Thermo Scientific™ NanoDrop™ One/OneC Security Suite Compliance Software
Shop All Thermo Scientific ProductsDescription
Confidently secure your laboratory data and more easily achieve compliance with US FDA 21 CFR Part 11 with Thermo Scientific™ Security Suite Software for use with the NanoDrop™ One/ OneC Microvolume Spectrophotometer. The software helps to facilitate compliant results in regulated environments with advanced tools such as user account access control, electronic signatures, audit trails, automated data exports, and security policies. Security Suite software integrates directly into the NanoDrop One PC Control Software for streamlined operation.
Security Suite for NanoDrop One/OneC
Software is designed to integrate into the free NanoDrop One/OneC PC control software.
Suite includes:
- Thermo Scientific Security Administration and Thermo Scientific Audit Manager
These two pieces of software enable users to run the NanoDropOne/OneC PC control software in a lab that needs to comply with FDA 21 CFR Part 11.
Software features include:
- User account access controlled through Windows log-in
- Electronic signaturesAudit trail
- Automated data export
- Security policies (example: prevent data deletion)
Included with the order:
- USB flash drive containing the Security Suite for the NanoDrop One/OneC PC control software installer
Specifications
Specifications
| Certifications/Compliance | 21 CFR Part 11 compliance |
| Display | Digital |
| Type | Security Suite for ND One/OneC Spectrophotometer |
| For Use With (Equipment) | NanoDrop One/OneC Spectrophotometers |
| Includes | Thermo Scientific Security Suite Software |