Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NanoDrop™ OneC Spectrophotometer & Qubit™ 4 Fluorometer with WiFi

Catalog No. A38190
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A38190 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A38190 Supplier Invitrogen™ Supplier No. A38190

For DNA, RNA or protein samples that are 100% pure, the results obtained on the Thermo Scientific™ NanoDrop™ OneC Microvolume UV-Vis Spectrophotometer and the Invitrogen™ Qubit™ 4 Fluorometer would be identical for samples within their dynamic ranges.

For DNA, RNA or protein samples that are 100% pure, the results obtained on the Thermo Scientific™ NanoDrop™ OneC Microvolume UV-Vis Spectrophotometer and the Invitrogen™ Qubit™ 4 Fluorometer would be identical for samples within their dynamic ranges. However, few samples are 100% pure.

As a bundled offering, the NanoDrop OneC Microvolume UV-Vis Spectrophotometer and the Qubit 4 Fluorometer together allow you to obtain the most complete information about the concentration and quality of your DNA, RNA or protein samples—helping to prevent costly troubleshooting downstream. While both the NanoDrop OneC spectrophotometer and the Qubit 4 fluorometer offer accurate measurements of DNA, RNA and protein samples, each also has its unique advantages.

NanoDrop OneC Spectrophotometer

Cited in more than 30,000 publications, the NanoDrop UV-Vis microvolume spectrophotometers have fundamentally changed the way scientists evaluate nucleic acid and protein samples by minimizing the need for large sample volumes.

The next generation NanoDrop OneC Microvolume UV-Vis Spectrophotometer utilizes innovative Thermo Scientific Acclaro™ Sample Intelligence technology that identifies contaminants, provides corrected concentration values and helps you understand the quality of your sample before you use it in downstream applications.

Qubit 4 Fluorometer

The Qubit 4 Fluorometer is a cutting-edge benchtop device that accurately measures DNA, RNA, and protein quantity, and now also RNA integrity and quality, using the highly sensitive Qubit assays.

For all Qubit assays, the concentration or quality of the target molecule in the sample is reported by a fluorescent dye that emits a signal only when bound to the target, which minimizes the effects of contaminants on the result. The easy-to-use touchscreen menus make it easy to select and run the assays you need, with results displayed in just a few seconds.

Comparison of the NanoDrop OneC spectrophotometer and Qubit 4 fluorometer

NanoDrop OneC Spectrophotometer

Qubit 4 Fluorometer

Fast and simple. Allows for direct measurement of purified samples with no dilutions required.

Overview

Sensitive and specific. Accurately measures concentration whether dsDNA, ssDNA, total RNA, microRNA or protein— even in the presence of contaminants.

UV absorbance

Technology

Fluorescence

Pipette sample onto pedestal; measure.

Workflow

Mix sample with reagents, incubate for 2–5 minutes, read.

Routine quantitation and qualification of DNA, RNA and proteins

Applications

Routine samples and with very dilute or difficult to process samples, when higher measurement sensitivity is required or when nucleic acid contaminants are suspected (e.g., DNA in RNA, dNTPs)

RNA sample quality

0.21–27,500

dsDNA quantitation range (ng/ul)

0.01–1,000

Acclaro Sample Intelligence technology identifies presence of contaminants such as protein or guanidine; provides corrected analyte concentration value.

Sample qualification (purity)

Unique fluorescent dyes enable differentiation between nucleic acid types (e.g., DNA vs. RNA) without any sample pre-treatment.

RNA IQ assay enables fast and facile detection of intact vs degraded in <5 seconds per sample

Selection of pre-programmed applications

  • Direct A280 quantitation with Protein Editor
  • A205 peptide bond absorbance
  • Colorimetric protein assays: Bradford, BCA, Lowry and Thermo Scientific Pierce™ 660 nm assay

Protein quantitation

Qubit assays exhibit minimal protein-to-protein variability and are resistant to most contaminants, including salts, reducing agents (e.g., DTT, b-mercaoptoethanol), DNA or amino acids, but not detergents.

1dsDNA quantitation range for NanoDrop OneC is 0.2 ng/μL with cuvette and 2.0 ng/μL without.

Key instrument features include:

NanoDrop OneC Microvolume UV-Vis Spectrophotometer

  • Fast and easy sample evaluation of nucleic acids and proteins with 'pipette, measure, clean' workflow and no sample carryover
  • Modern stand-alone design with local control and high-resolution touchscreen interface, which saves bench space
  • Accurate measurements up to 27,500 ng/μL (dsDNA) with extended auto-range path length technology
  • No need for cuvettes or consumables needed so you can pipette directly onto pedestal sample retention system
  • Pre-programmed applications designed specifically for life scientists
  • Enhanced connectivity with data transfer via USB, Ethernet, Bluetooth™ and WiFi options; PC software available for data management
  • Cuvette position for measuring dilute solutions and performing temperature sensitive experiments with the NanoDrop OneC spectrophotometer

Qubit 4 Fluorometer

  • Fast and highly accurate quantitation of DNA, RNA, and protein in less than three seconds per sample
  • Measures intact RNA in less than 5 seconds per sample
  • High levels of accuracy using only 1–20 μL of sample, even with very dilute samples
  • On-board Reagent Calculator quickly generates Qubit working solution preparation instructions
  • Stores results from up to 1000 samples
  • Large 5.7-inch, state-of-the-art color touchscreen for easy workflow navigation
  • Graphical display indicates when samples are in the extended range or out of range
  • Small footprint saves space on your bench
  • Flexibility to export data to using the included WiFi dongle, a USB drive, or directly to your computer via a USB cable
  • Ability to personalize your Qubit fluorometer with the assays you run most, add new assays, or even create your own assays with the MyQubit software and web tool
  • Language of your choice including English, French, Spanish, Italian, German, simplified Chinese, and Japanese
TRUSTED_SUSTAINABILITY

Specifications

Display Touchscreen
Product Line NanoDrop Qubit
For Use With (Application) Quantitation and qualification of DNA, RNA and proteins
Quantity 1 each
Sample Volume (Metric) 0.21 μL
Storage Requirements Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.