Learn More
Thermo Scientific™ NanoDrop™ Onec Spectrophotometer with WiFi and Invitrogen Qubit™ 4 Fluorometer
Shop All Thermo Scientific ProductsDescription
- NanoDrop Onec Microvolume UV-Vis Spectrophotometer
- Fast and easy sample evaluation of nucleic acids and proteins with pipette, measure, clean workflow and no sample carryover
- Modern stand-alone design with local control and high-resolution touchscreen interface, which saves bench space
- Accurate measurements up to 27500 ng/μL (dsDNA) with extended auto-range path length technology
- No need for cuvettes or consumables needed so you can pipette directly onto pedestal sample retention system
- Pre-programmed applications designed specifically for life scientists
- Enhanced connectivity with data transfer via USB, Ethernet, Bluetooth™ and WiFi options; PC software available for data management
- Cuvette position for measuring dilute solutions and performing temperature sensitive experiments with the NanoDrop OneC spectrophotometer
- Qubit 4 Fluorometer
- Fast and highly accurate quantitation of DNA, RNA, and protein in less than three seconds per sample
- Measures intact RNA in less than 5 seconds per sample
- High levels of accuracy using only 1 to 20 μL of sample, even with very dilute samples
- On-board Reagent Calculator quickly generates Qubit working solution preparation instructions
- Stores results from up to 1000 samples
- Large 5.7-inch, state-of-the-art color touchscreen for easy workflow navigation
- Graphical display indicates when samples are in the extended range or out of range
- Small footprint saves space on your bench
- Exports data to a USB drive or directly to your computer via a USB cable
- Ability to personalize your Qubit fluorometer with the assays you run most, add new assays, or even create your own assays with the MyQubit software and web tool
- Language of your choice including English, French, Spanish, Italian, German, simplified Chinese, and Japanese
- Dynamic Range: 5 orders of magnitude
- Memory: 4 GB USB flash drive
- Emission filters: green (510 to 580 nm), red (665 to 720 nm)
- Excitation filters: blue (430 to 495 nm), red (600 to 645 nm)
- LED light sources: blue (max 470 nm), red (max 635 nm)
- Photodiode detectors: 300 to 1000 nm range
Specifications
Specifications
| Certifications/Compliance | CE and UL/CSA Approved |
| Compatibility | DYMO LabelWriter 450 printer, Bluetooth keyboard, mouse and barcode reader |
| Concentration | dsDNA: Pedestal 27500 ng/μL; BSA (IgG): Pedestal 820 (400) mg/mL |
| Connections | Three USB-A ports, Ethernet, Bluetooth™, and Wi-Fi |
| Detection Range | dsDNA: Pedestal 2 ng/μL, Cuvette 0.2 ng/μL; BSA (IgG): Pedestal 0.06 (0.03) mg/mL, Cuvette 0.006 (0.003) mg/mL |
| Detector Type | 2048-element CMOS Linear Image Sensor |
| Display | 7 in. High Definition Color Display, 1280 x 800 pixels |
| Dimensions (L x W x H) | 20 x 25.4 x 32.3 cm (8 x 10 x 12.7 in.) |
| Languages | English, French, German, Spanish, Polish, Chinese, Japanese, Korean |
| Light Source | Xenon Flash |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.