Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NanoDrop™ Reagents

Catalog No. 13400515 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
CF-1 Calibration Check Fluid
CF-8 Calibration Check Kit
ND PR-1 Reconditioning Kit
PV-1 Performance Verification
PV-8 Performance Verification
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-400-515 PV-1 Performance Verification
CF1 CF-1 Calibration Check Fluid
CF8 CF-8 Calibration Check Kit
PR1KIT ND PR-1 Reconditioning Kit
13-400-532 PV-8 Performance Verification
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13-400-515 Supplier Thermo Scientific™ Supplier No. CHEMPV1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified standards for verifying instrument performance to factory specifications are available separately.

Features include:
  • Solution with certified absorbance values
  • Use to create a record of correct photometric performance of a NanoDrop One spectrophotometer
TRUSTED_SUSTAINABILITY

Specifications

Type PV-1 Performance Verification
For Use With (Equipment) NanoDrop Ultra, NanoDrop One and NanoDrop OneC spectrophotometers
Storage Requirements Store at room temperature
Can I verify that my NanoDrop One/Onec Spectrophotometer is calibrated and running accurately?

Yes. While most users will never need to recalibrate their NanoDrop One/Onec Spectrophotometer, we recommend verifying instrument performance using the NanoDrop PV-1 Performance Verification Solution every 6 months. PV-1 is available from the Thermo Fisher website.

WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.