Learn More
Thermo Scientific NanoDrop™ Reagents
Catalog No. PR1KIT Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
CF-1 Calibration Check Fluid
CF-8 Calibration Check Kit
ND PR-1 Reconditioning Kit
PV-1 Performance Verification
PV-8 Performance Verification
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
PR1KIT ND PR-1 Reconditioning Kit
CF1 CF-1 Calibration Check Fluid
CF8 CF-8 Calibration Check Kit
13-400-515 PV-1 Performance Verification
13-400-532 PV-8 Performance Verification
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PR1KIT Supplier Thermo Scientific Supplier No. CHEMPR1KIT
Add to Cart
Add to Cart
Certified standards for verifying instrument performance to factory specifications are available separately.
This kit is useful for reconditioning the sample measurement pedestals to a hydrophobic state if they become “unconditioned” (see user manual for more information). The PR-1 kit consists of a small screw cap vial of specially formulated compound and a supply of 25 convenient applicators. Kit sold separately from instrument.
For use with Thermo Scientific™ NanoDrop™ Spectrophotometers.
Includes:
Small screw cap vial of specially formulated reconditioning compound and 25 convenient swabs
TRUSTED_SUSTAINABILITY
Specifications
Type ND PR-1 Reconditioning Kit
For Use With (Equipment) All Thermo Scientific NanoDrop Spectrophotometers
How do I keep my sample from spreading on the pedestal of my NanoDrop Lite Spectrophotometer?
There are many reagents and organic solvents that could compromise the pedestal surface properties, causing samples to flatten out rather than bead up. Use the NanoDrop PR-1 reconditioning compound as a rapid means of reconditioning the pedestals when the pedestal surface properties have been compromised and samples spread out on the pedestal. NanoDrop PR-1 Reconditioning Kits are available through Thermo Fisher website (Cat. No. CHEM-PR1-KIT)
My NanoDrop 8000 Spectrophotometer failed calibration check. What should I do?
Please review the following steps to take when/if your NanoDrop 8000 Spectrophotometer fails a calibration check:
- Clean the pedestal with the NanoDrop PR-1 Reconditioning Kit
- Repeat calibration check using a FRESH vial of NanoDrop CF-1 Calibration Fluid and FRESH blanking solution
- If instrument continues to be out of specification, Contact Technical Support
My NanoDrop 2000/2000c Spectrophotometer failed the calibration check. What should I do?
Please review the following steps to take when/if your NanoDrop 2000/2000c fails a calibration check:
- Clean the pedestal with the NanoDrop PR-1 Reconditioning Kit
- Repeat calibration check using a FRESH vial of NanoDrop CF-1 Calibration Fluid and FRESH blanking solution
More detailed instructions can be found on the Calibration Check Procedure document: https://assets.thermofisher.com/TFS-Assets/CAD/Product-Bulletins/NanoDrop-2000-2000c-1000-Calibration-Check-procedure.pdf
If your NanoDrop 2000/2000c continues to be out of specification, Contact Technical Support.
WARNING: Cancer - www.P65Warnings.ca.gov