Learn More
Thermo Scientific™ NanoDrop™ Ultra FL-1 Fluorescence Verification Kit
Shop All Thermo Scientific ProductsDescription
The Thermo Scientific NanoDrop Ultra FL-1 Fluorescence Verification Kit is a specialized assay system designed to ensure optimal performance verification of fluorometer components in NanoDrop Ultra FL and UltraC FL spectrophotometers. This comprehensive kit provides essential reagents, including Blank Reagent, Green Fluorescence Reagent, and Far Red Fluorescence Reagent.
Engineered specifically for researchers conducting RNA analyses through qPCR and RT-PCR methodologies, the NanoDrop Ultra FL-1 Fluorescence Verification Kit supports the advanced microliter multimode detection capabilities of NanoDrop Ultra/UltraC FL instruments. The integration of robust fluorescence assay verification ensures streamlined workflows while maintaining effortless compliance with 21 CFR Part 11 regulatory requirements.
Features of the NanoDrop Ultra FL-1 Fluorescence Verification Kit include:
- Comprehensive reagent kit—contains three essential reagents, including Blank Reagent, Green Fluorescence Reagent, and Far Red Fluorescence Reagent for complete system calibration and performance verification
- Instrument-specific compatibility—engineered exclusively for NanoDrop Ultra FL and UltraC FL spectrofluorometer models to ensure optimal system integration and measurement accuracy
- User-friendly implementation—supplied with comprehensive setup and operational instructions to facilitate proper reagent handling and calibration procedures
- Preventive maintenance protocol—designed for biannual application (every six months) to maintain instrument precision and extend operational lifespan
- Multi-wavelength calibration capability—enables calibration across multiple fluorescence detection channels through green and far-red fluorescent standards
- Quality assurance standards—provides blank reference material for baseline correction and systematic error identification during routine instrument validation
Laboratory quality assurance and measurement integrity
By implementing the NanoDrop Ultra FL-1 Fluorescence Verification Assay Kit, laboratories establish a robust quality assurance framework that ensures their NanoDrop Ultra FL and UltraC FL devices maintain optimal functional performance. This systematic approach to instrument validation directly supports the maintenance of measurement integrity across all fluorescence-based analytical procedures. The verification process enables laboratories to demonstrate compliance with quality standards while providing documented evidence of instrument performance, thereby supporting regulatory requirements and internal quality management systems.
Warranty and Services
3 Year Warranty
Specifications
Specifications
| Assay | Fluorescence Verification |
| Content And Storage | 2°C to 8°C |
| Detection Method | Fluorescence |
| Format | Kit |
| For Use With (Equipment) | NanoDrop Ultra |
| No. of Reactions | 25 Assays |
| Product Type | Verification Kit |
| Quantity | 1 Kit |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.