Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific NanoDrop Ultra FL Acclaro Pro Security and IQOQ Bundle

Catalog No. 13400546 Shop All Thermo Scientific Products
Change view
Click to view available options
Spectral Bandwidth:
≤1.8 nm (FWHM at Hg 254 nm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Spectral Bandwidth
13-400-546 ≤1.8 nm (FWHM at Hg 254 nm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-400-546 Supplier Thermo Scientific Supplier No. NDUFLAPSECUS
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Helps deliver fast, accurate microvolume UV-Vis and fluorescence analysis with Acclaro Pro, SciVault 2, and IQOQ Kit.

  • The Ultra Spectrophotometers and Fluorometers represent the next generation of microvolume detection with exceptional accuracy and sensitivity for nucleic acid and protein analysis using as little as 1.0 μL.
  • Designed to integrate seamlessly into workflows across genomics, proteomics, molecular biology, biotechnology, pharmaceutical, and life sciences research.
  • Delivers unparalleled precision, efficiency, and reliability in one compact device for consistent microvolume performance.
  • Bundles include one NanoDrop Ultra instrument with additional factory calibration unlocking Thermo Scientific™ Acclaro™ Pro software, SciVault™ 2 activation, and a model-specific IQ/OQ kit.
  • Thermo Scientific™ Acclaro™ Sample Intelligence technology enhances measurement accuracy and identifies contaminants for reliable data integrity.
  • Quantify and qualify DNA, RNA, and protein samples in seconds using only 1 to 2 μL, with full spectral data for downstream decisions.
  • Cuvette position available for additional flexibility during measurements.
  • Optional Thermo Scientific™ SciVault™ 2 software provides tools for 21 CFR Part 11 compliance through user account management, digital signatures, and audit logs.
  • Integrates with NanoDrop Ultra software and can operate on-board or via a connected domain for centralized or local control.
  • Centralized SciVault 2 setup enables control of user privileges and audit logs across multiple NanoDrop Ultra instruments in different labs.
  • Fully integrated qPCR recipe calculator simplifies reaction setup by automatically computing volumes based on sample concentration.
  • Auto-Measure and Auto-Blank functions streamline multi-sample workflows with instant measurements when the pedestal arm is lowered; toggle ON or OFF with a single tap.
  • Export results directly to cloud storage platforms like Microsoft OneDrive, Google Drive, or Thermo Fisher Connect.
  • Direct LIMS integration allows seamless data transfer using the built-in Restful API.
  • Print results directly from the NanoDrop Ultra instrument via Wi-Fi, Ethernet, or USB, or print exported data using NanoDrop Ultra PC Control software.
  • Customize and preview exported data content to match downstream requirements before saving or sharing.
  • Acclaro Pro Software delivers ±5% error accuracy across the absorbance dynamic range, providing unmatched precision.
  • Measure high concentrations accurately - up to 550 Abs (10 mm equivalent), 400 mg/mL IgG, or 18,000 ng/μL with dilution-free workflows.
  • Perform triplicate measurements on the pedestal in under two minutes for quick, high-accuracy analysis.
  • Enhanced calibration from the factory ensures exceptional accuracy within Acclaro Pro applications.
  • Model-specific IQ/OQ Kits include USB flash drive with IQ/OQ procedure, CHEM-PR1-KIT, CHEM-PV-1, and a Cuvette Qualification Kit (840-257300) if a cuvette model is purchased; CHEM-FL-1 for fluorescence models.
  • For research use only; not for diagnostic purposes.
TRUSTED_SUSTAINABILITY

Specifications

Detector Type 2048-element CMOS Linear Image Sensor
Display Color Touchscreen
Languages English, Chinese, French, Japanese, German, Spanish, Polish, Italian, Korean
Light Source Xenon Flash, LED
Product Line NanoDrop
Spectral Bandwidth ≤1.8 nm (FWHM at Hg 254 nm)
Type Spectrophotometers Bundle
Voltage 100/240 VAC
Wavelength Range 190 to 850 nm
Wavelength Selection Automatic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.