Learn More
Thermo Scientific™ NanoDrop Ultra IQ/OQ Kits
Catalog No. 13400600
Change view
Click to view available options
Type:
ND UltraC FL + IQ/OQ
NanoDrop FL and IQ/OQ Kit
NanoDrop Ultra and IQ/OQ Kit
NanoDrop UltraC and IQ/OQ Kit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-400-600 NanoDrop Ultra and IQ/OQ Kit
13-400-601 NanoDrop UltraC and IQ/OQ Kit
13-400-540 NanoDrop FL and IQ/OQ Kit
13-400-539 ND UltraC FL + IQ/OQ
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-400-600 Supplier Thermo Scientific™ Supplier No. NDUIQOQKIT
Add to Cart
Add to Cart
Purchase a Thermo Scientific™ NanoDrop™ Ultra IQ/OQ Kit to receive all the model-specific documentation and reference materials needed to faciliate Installation and Operational qualification procedures.
Purchase a Thermo Scientific™ NanoDrop™ Ultra IQ/OQ Kit to receive all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ). Certified standards for verifying instrument performance to factory specifications are available separately.
UV-Visible Spectrophotometer and Fluorometer Validation Materials include:
  • Information on the validation process
  • Documentation for manufacturer-verified Installation Qualification (IQ) to ensure proper system performance
  • Installation and instrument certification
NanoDrop Ultra IQOQ Kit
Includes:
  • USB flash drive containing IQ/OQ procedure for NanoDrop Ultra
  • CHEM-PR1-KIT
  • CHEM-PV-1
NanoDrop UltraC IQOQ Kit
Includes:
  • USB flash drive containing IQ/OQ procedure for NanoDrop Ultra
  • CHEM-PR1-KIT
  • CHEM-PV-1
  • Cuvette Qualification Kit (840-257300)
Proper application of these materials ensures your instrument is installed correctly and working within manufacturer's specifications.
TRUSTED_SUSTAINABILITY
Specifications
Product Line NanoDrop
Type NanoDrop Ultra and IQ/OQ Kit
For Use With (Equipment) NanoDrop Ultra Spectrophotometers and Fluorometers
Includes Documentation and materials to facilitate installation and operational qualification procedures (IQ/OQ) for use with NanoDrop Ultra Spectrophotometers and Fluorometers