Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NanoDrop Ultra and Qubit 4 Bundle

Catalog No. 13622610 Shop All Thermo Scientific Products
Change view
Click to view available options
Light Source:
Xenon Flash
Display:
10 in. High Definition Color Display
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Light Source Display
13-622-610 Xenon Flash 10 in. High Definition Color Display
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-622-610 Supplier Thermo Scientific™ Supplier No. NDULTRAQB4US
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Combining the NanoDrop Ultra and Qubit 4 allows for a more comprehensive analysis. Using both tools allows researchers to quickly screen samples with NanoDrop Ultra and then perform more accurate quantification on selected samples using Qubit 4

Accuracy and Sensitivity:

  • NanoDrop Ultra: Provides quick and easy quantification of nucleic acids and proteins directly from a small sample volume without the need for reagents. However, it may be less sensitive for low-concentration samples.
  • Qubit 4: Uses fluorescent dyes specific to the target molecule, offering high sensitivity and accuracy for low-concentration samples.

Complementary Data:

  • NanoDrop Ultra: Offers absorbance-based quantification, which is useful for a broad range of concentrations but in most cases required the sample to be purified. Acclaro contamination alerts provide the user with information on possible contaminats in the sample.
  • Qubit 4: Provides fluorescence-based quantification, which is less affected by contaminants and more specific for the target molecule.

Verification:

  • Using both instruments can help verify results. If the NanoDrop and Qubit measurements agree, it increases confidence in the data.
  • Discrepancies might indicate the presence of contaminants or other issues that need to be addressed.

Efficiency:

  • Combining both tools allows researchers to quickly screen samples with NanoDrop Ultra and then perform more accurate quantification on selected samples using Qubit 4.

Broader Application:

  • NanoDrop Ultra: Suitable for routine measurements and quick checks.
  • Qubit 4: Ideal for precise quantification needed in downstream applications like qPCR, sequencing, or other sensitive assays.
TRUSTED_SUSTAINABILITY

Specifications

Detector Type 2048-element CMOS Linear Image Sensor
Display 10 in. High Definition Color Display
Languages English, Chinese, Italian, Spanish, Japanese
Light Source Xenon Flash
Product Line NanoDrop
Spectral Bandwidth 1.8 nm
Type NanoDrop Ultra and Qubit 4 Bundle
Voltage 12 VDC
Wavelength Range 190 to 850 nm
Wavelength Selection Automatic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.