Learn More
Thermo Scientific™ NanoDrop Ultra SciVault 2 Compliance Software
Catalog No. 13400620 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
US FDA 21 CFR Part 11 Compliance Software
Catalog No. Type
13-400-620 US FDA 21 CFR Part 11 Compliance Software
1 options
Catalog No. 13-400-620 Supplier Thermo Scientific™ Supplier No. NDUSCIVAULT2
Add to Cart
Add to Cart
Purchase the Thermo Scientific™ SciVault™ 2 software, to be used with any NanoDrop™ Ultra model, which enables 21 CFR Part 11 compliance and keeps electronic records secure.
Confidently secure your laboratory data and more easily achieve compliance with US FDA 21 CFR Part 11 with Thermo Scientific™ SciVault™ 2 Software for use with the NanoDrop™ Ultra Microvolume Spectrophotometers and Fluorometers. The software helps to facilitate compliant results in regulated environments with advanced tools such as user account management, user account access control, electronic signatures, audit trails, automated data exports, and security policies. This SciVault 2 software is available for both on-board and PC controlled software applications.
Tools to aid in 21 CFR Part 11 compliance
Optional Thermo Scientific™ SciVault™ 2 software can be activated through the local on-board software or installed on a companion PC and gives labs the tools they need to comply with US FDA 21 CFR Part 11.
  • Compliance made easy – SciVault 2 integrates into the NanoDrop Ultra software user interface and controls user account management, user account access, digital signatures, and audit logs.
User Management feature: Create users or pull existing users from a connected active directory and assign them to unique roles.
  • User Privileges feature: This policy setting feature allows policies to be assigned to each role. Policies determine what access and restrictions are assigned to each role.
  • Audit Logs feature: Generate and save an audit trail of activities that have occurred related to the instrument and software.
  • On-board options — Software is available to be run directly on the local instrument through on–board user account management or you can connect your instrument to your domain to use your Windows account management.
  • Centralized solution - install SciVault 2 software on a central computer to control privileges and view audit logs across multiple NanoDrop Ultra instruments in different labs.
Purchase includes:
USB stick containing the SciVault 2 software, which can be installed on a companion PC, and a Key file that can be used to unlock SciVault 2 on the NanoDrop Ultra instrument itself.
TRUSTED_SUSTAINABILITY
Specifications
Type US FDA 21 CFR Part 11 Compliance Software
For Use With (Equipment) NanoDrop Ultra Spectrophotometers and Fluorometers
Includes USB flash drive containing the SciVault 2 software for use with the NanoDrop Ultra Spectrophotometers and Fluorometers