Learn More
Thermo Scientific™ NanoDrop Ultra USB WiFi and Bluetooth Dongle
Catalog No. 13400614 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
NanoDrop Ultra USB WiFi and Bluetooth Dongle
Catalog No. Includes
13-400-614 NanoDrop Ultra USB WiFi and Bluetooth Dongle
1 options
Catalog No. 13-400-614 Supplier Thermo Scientific™ Supplier No. NDUWIFISTICK
Add to Cart
Add to Cart
Purchase a NanoDrop™ Ultra USB WiFi and Bluetooth Dongle to enable Wi-Fi and Bluetooth connectivity to NanoDrop Ultra UV-Vis Spectrophotometers and Fluorometers.
Upgrade your wireless-enabled Thermo Scientific™ NanoDrop™ Ultra Spectrophotometer and Fluorometer with this high-power USB Wi-Fi and Bluetooth Dongle. Achieve high speed transfer rates and maximum range of connectivity during remote monitoring of your instrument.
Key features and benefits:
  • Leading wireless technology standard for top performance and range
  • Maximum speed ofup to 300 Mbps for fast wireless transfer rates
  • Extensive range with built-in high-power antenna
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) NanoDrop Ultra Spectrophotometers and Fluorometer
Includes NanoDrop Ultra USB WiFi and Bluetooth Dongle