Learn More
Thermo Scientific NanoDrop UltraC FL Acclaro Pro Security and IQOQ Bundle
Shop All Thermo Scientific ProductsDescription
- The Ultra Spectrophotometers and Fluorometers represent the next generation of microvolume detection, offering exceptional accuracy and sensitivity for nucleic acid and protein analysis with sample volumes as low as 1.0 μL.
- Engineered to integrate seamlessly into workflows across genomics, proteomics, molecular biology, biotechnology, pharmaceutical, and life sciences research applications.
- Delivers precision, efficiency, and reliability in one compact design, ideal for microvolume analysis of DNA, RNA, and protein samples.
- Each bundle includes a NanoDrop Ultra instrument with an additional factory calibration unlocking Thermo Scientific™ Acclaro™ Pro software, Thermo Scientific™ SciVault™ 2 activation, and a model-specific IQ/OQ kit.
- Thermo Scientific™ Acclaro™ Sample Intelligence technology enhances measurement accuracy and identifies contaminants for improved data reliability.
- Quantify and qualify DNA, RNA, and protein samples in seconds using only 1 to 2 μL, obtaining full spectral data before proceeding with downstream applications.
- Includes a cuvette position for additional measurement flexibility across different sample types.
- Thermo Scientific™ SciVault™ 2 software supports laboratories in meeting US FDA 21 CFR Part 11 compliance through user management, audit trails, and secure data logging.
- Integrates directly with NanoDrop Ultra software for easy management of user accounts, access controls, digital signatures, and audit logs.
- Can be configured for local on-board control or connected to a networked domain for centralized user authentication and audit log management.
- Centralized SciVault 2 setup allows monitoring and managing privileges and audit logs across multiple NanoDrop Ultra instruments in different labs.
- Fully integrated qPCR recipe calculator automates reaction setup using preloaded or custom kit libraries to simplify sample preparation.
- Auto-Measure and Auto-Blank functions streamline multi-sample workflows with instant measurements when the pedestal arm is lowered; functions can be toggled ON or OFF via the touchscreen interface.
- Advanced connectivity enables direct data export to cloud platforms such as Microsoft OneDrive, Google Drive, and Thermo Fisher Connect.
- Direct LIMS integration allows data transfer via an integrated Restful API for efficient digital record management.
- Data-to-printer options include Wi-Fi, Ethernet, or USB printing directly from the instrument, or through NanoDrop Ultra PC Control software.
- Customize and preview export data content to ensure it meets regulatory, reporting, or downstream workflow needs.
- Acclaro Pro software delivers ±5% error across the absorbance dynamic range for accurate high-concentration measurement performance.
- Supports absorbance up to 550 Abs (10 mm equivalent), up to 400 mg/mL IgG or 18,000 ng/μL for precise, dilution-free quantification.
- Performs triplicate measurements on the pedestal in under two minutes for rapid, reproducible results.
- Enhanced factory calibration ensures superior accuracy within Acclaro Pro applications for consistent results.
- Model-specific IQ/OQ Kits include a USB flash drive with IQ/OQ procedures, CHEM-PR1-KIT, CHEM-PV-1, and a Cuvette Qualification Kit (840-257300) for cuvette models or CHEM-FL-1 for fluorescence models.
- For research use only; not for diagnostic purposes.
Specifications
Specifications
| Detector Type | 2048-element CMOS Linear Image Sensor |
| Display | Color Touchscreen |
| Languages | English, Chinese, French, Japanese, German, Spanish, Polish, Italian, Korean |
| Light Source | Xenon Flash, LED |
| Product Line | NanoDrop |
| Spectral Bandwidth | ≤1.8 nm (FWHM at Hg 254 nm) |
| Type | Spectrophotometers Bundle |
| Voltage | 100/240 VAC |
| Wavelength Range | 190 to 850 nm |
| Wavelength Selection | Automatic |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.