Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific NanoDrop UltraC FL dsDNA & RNA BR Fluorescence Assay Bundle

Catalog No. 13400547 Shop All Thermo Scientific Products
Change view
Click to view available options
Spectral Bandwidth:
≤1.8 nm (FWHM at Hg 254 nm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Spectral Bandwidth
13-400-547 ≤1.8 nm (FWHM at Hg 254 nm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-400-547 Supplier Thermo Scientific Supplier No. NDUFLD/RBNDLUS
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Combines spectrophotometer and fluorometer with dsDNA and RNA BR Assay Kits for specific and accurate quantification.

  • Avoid costly setbacks in your workflow with an understanding of sample quality like never before using the Thermo Scientific™ NanoDrop™ Ultra Microvolume UV-Vis Spectrophotometers Fluorometers.
  • Innovative Thermo Scientific™ Acclaro™ Sample Intelligence technology is built into every instrument for improved measurement accuracy and contaminant identification.
  • Quantify and qualify DNA, RNA, and protein samples in seconds with only 1 to 2 μL, and obtain full-spectral data before you decide to use samples in downstream applications.
  • Additional cuvette position also available for greater flexibility in experimental workflows.
  • Each bundle includes a NanoDrop™ Ultra fluorescence-capable instrument (NanoDrop Ultra FL or NanoDrop UltraC FL) with both the NanoDrop Ultra dsDNA BR and RNA BR Assay Kits.
  • Obtain comprehensive information about the concentration and quality of DNA or RNA samples through absorbance and fluorescence measurements on the same instrument.
  • The NanoDrop Ultra FL and NanoDrop UltraC FL perform both micro-volume absorbance and fluorescence measurements using red and blue solid-state LEDs.
  • High Specificity - Fluorescence offers greater selectivity than absorbance-based quantification, allowing accurate detection even in impure or complex samples.
  • Sample Prep Calculator - integrated reagent calculator determines the required amount of dye and buffer for each assay.
  • Fully integrated qPCR recipe calculator - use the built-in library of kits or add your own to automate reaction preparation based on sample concentration.
  • Auto-Measure and Auto-Blank functions streamline multi-sample processing with instant measurements triggered when the pedestal arm is lowered; both functions can be toggled ON or OFF with a single tap.
  • Export to the cloud - transfer data directly to Microsoft OneDrive, Google Drive, or Thermo Fisher Connect for secure cloud storage.
  • Direct LIMS Integration - send data directly to your laboratory information management system using the built-in Restful API.
  • Flexible data printing options - print directly from the instrument via Wi-Fi, Ethernet, or USB, or print exported data through NanoDrop Ultra PC Control software.
  • Data Your Way - customize and preview data file content before exporting to ensure compatibility with downstream applications.
  • For research use only; not for diagnostic purposes.
TRUSTED_SUSTAINABILITY

Specifications

Detector Type 2048-element CMOS Linear Image Sensor
Display Color Touchscreen
Languages English, Chinese, French, Japanese, German, Spanish, Polish, Italian, Korean
Light Source Xenon Flash, LED
Product Line NanoDrop
Spectral Bandwidth ≤1.8 nm (FWHM at Hg 254 nm)
Type Spectrophotometers Bundle
Voltage 100/240 VAC
Wavelength Range 190 to 850 nm
Wavelength Selection Automatic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.