Learn More
Waters Corp nanoEase M/Z HSS T3 Column, 100Ã…, 1.8 µm, 300 µm X 50 mm, 1/pk;
Supplier:  Waters Corp 186009252
Encompass_Preferred
nanoEase M/Z HSS T3 Column, 100Å, 1.8 µm, 300 µm X 50 mm, 1/pk;
Catalog No. 50-203-8947