Learn More
Waters Corp nanoEase M/Z Peptide BEH C18 Column, 300Å, 1.7 μm, 75 μm × 200 mm
Catalog No. 502391100 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This nanoEase M/Z Peptide BEH C18 Column supports high-performance nano- to micro-scale liquid chromatographic separations, providing excellent sensitivity, resolution, and reproducibility for biomarker discovery and protein characterization.
  • Optimal performance for nano- to micro-scale liquid chromatography.
  • High sensitivity, resolution, and reproducibility for biomarker discovery.
  • Withstands 15kpsi operating pressure for superior longevity.
  • Patented BEH Technology with 300Å pore, C18, 1.7 μm particles.
  • Wide-pore (300Å) BEH particle: broad pH range, low pH stability, ultra-low column bleed.
  • QC tested with gradient separation of cytochrome c tryptic digest.
  • ZenFit connector for easy, fingertight, leak-free liquid connections.
  • Fittings compatible with V-Detail ports and Rheodyne/Vici valves.
Catalog No. 50-239-1100 Supplier Waters Corp Supplier No. 186008800
This nanoEase M/Z Peptide BEH C18 Column supports high-performance nano- to micro-scale liquid chromatographic separations, providing excellent sensitivity, resolution, and reproducibility for biomarker discovery and protein characterization.
  • Optimal performance for nano- to micro-scale liquid chromatography.
  • High sensitivity, resolution, and reproducibility for biomarker discovery.
  • Withstands 15kpsi operating pressure for superior longevity.
  • Patented BEH Technology with 300Å pore, C18, 1.7 μm particles.
  • Wide-pore (300Å) BEH particle: broad pH range, low pH stability, ultra-low column bleed.
  • QC tested with gradient separation of cytochrome c tryptic digest.
  • ZenFit connector for easy, fingertight, leak-free liquid connections.
  • Fittings compatible with V-Detail ports and Rheodyne/Vici valves.