Learn More
Waters Corp nanoEase M/Z Peptide BEH C18 Column, 130 Å, 1.7 μm, 100 μm x 100 mm
Catalog No. 502391487 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The nanoEase M/Z Peptide BEH C18 Column is designed for optimal performance in nano- to micro-scale liquid chromatographic separation. It provides high sensitivity, resolution, and reproducibility for biomarker discovery, protein identification, and characterization. These columns can withstand 15000 psi operating pressure and handle pressure fluctuations during injection, contributing to superior longevity.
  • Offers high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification.
  • Withstands 15000 psi operating pressure for superior longevity.
  • Contains BEH Technology 130 Å pore, C18, 1.7 μm particles.
  • Small-pore BEH particle offers widest usable pH range and superior low pH stability.
  • Equipped with ZenFit connectors for easy, fingertight, tool-free, leak-free connections.
  • Manufactured in a cGMP, ISO 9001-certified facility.
Catalog No. 50-239-1487 Supplier Waters Corp Supplier No. 186008796
The nanoEase M/Z Peptide BEH C18 Column is designed for optimal performance in nano- to micro-scale liquid chromatographic separation. It provides high sensitivity, resolution, and reproducibility for biomarker discovery, protein identification, and characterization. These columns can withstand 15000 psi operating pressure and handle pressure fluctuations during injection, contributing to superior longevity.
  • Offers high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification.
  • Withstands 15000 psi operating pressure for superior longevity.
  • Contains BEH Technology 130 Å pore, C18, 1.7 μm particles.
  • Small-pore BEH particle offers widest usable pH range and superior low pH stability.
  • Equipped with ZenFit connectors for easy, fingertight, tool-free, leak-free connections.
  • Manufactured in a cGMP, ISO 9001-certified facility.